Assay Details
Target Gene Details
Entrez Gene ID: | 52685 |
Gene Name: | CD300 antigen like family member G |
Gene Aliases: |
2310016B05Rik, Clm9, D11Ertd736e |
Location: |
Chr.11:102041511-102055617 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd300lg | NM_001160711.1 | NP_001154183.1 | ||
| NM_001160712.1 | NP_001154184.1 | |||
| NM_001160713.1 | NP_001154185.1 | |||
| NM_027987.3 | NP_082263.2 | |||
| AB243063.1 | BAE96046.1 | |||
| AB243064.1 | BAE96047.1 | |||
| AB243065.1 | BAE96048.1 | |||
| AB243066.1 | BAE96049.1 | |||
| AK009375.1 | BAB26251.1 | |||
| AK171982.1 | BAE42759.1 | |||
| AY457055.1 | AAR27946.1 | |||
| BC116906.1 | ||||
| BC119315.1 |
More Information
Set Membership: |
|

Genomic Map