Assay Details
Target Gene Details
Entrez Gene ID: | 15985 |
Gene Name: | CD79B antigen |
Gene Aliases: |
B29, Ig-beta, Igb, Igbeta |
Location: |
Chr.11:106311341-106314529 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd79b | NM_008339.2 | NP_032365.1 | ||
| AK143573.1 | BAE25444.1 | |||
| BC012226.1 | AAH12226.1 | |||
| CT010358.1 | CAJ18566.1 | |||
| J03857.1 | AAA37274.1 |
More Information
Set Membership: |
|

Genomic Map