Assay Details
Target Gene Details
Entrez Gene ID: | 217232 |
Gene Name: | cell division cycle 27 |
Gene Aliases: |
AI452358, APC3, BC023187 |
Location: |
Chr.11:104502524-104550620 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc27 | NM_001285988.1 | NP_001272917.1 | ||
| NM_001285989.1 | NP_001272918.1 | |||
| NM_001285990.1 | NP_001272919.1 | |||
| NM_145436.2 | NP_663411.2 | |||
| AK148278.1 | ||||
| AK161143.1 | BAE36213.1 | |||
| AK161471.1 | ||||
| BC023187.1 | 11 | 1416 | AAH23187.1 | |
| BC157955.1 | ||||
| BC172100.1 |
More Information
Set Membership: |
|

Genomic Map