Assay Details
Target Gene Details
Entrez Gene ID: | 67480 |
Gene Name: | CWC25 spliceosome-associated protein homolog (S. cerevisiae) |
Gene Aliases: |
1300013D05Rik, AA473636, Ccdc49, R75228 |
Location: |
Chr.11:97745470-97766613 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cwc25 | NM_026186.4 | 3 | 490 | NP_080462.2 |
| AK004983.2 | 3 | 434 | BAB23718.1 | |
| AK017849.1 | 3 | 435 | ||
| AK030628.1 | 3 | 419 | ||
| AK035158.1 | 3 | 419 | ||
| BC021478.1 | 3 | 404 | ||
| BC079553.1 | 3 | 412 | AAH79553.1 |

Genomic Map