Assay Details
Target Gene Details
Entrez Gene ID: | 72729 |
Gene Name: | CDC42 small effector 2 |
Gene Aliases: |
2810404F18Rik, AA408783, AA536669, SPEC2 |
Location: |
Chr.11:54717415-54787703 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42se2 | NM_178626.3 | NP_848741.1 | ||
| AK039301.1 | BAC30311.1 | |||
| AK080473.1 | BAC37927.1 | |||
| AK088090.1 | BAC40139.1 | |||
| AK090381.1 | BAC41193.1 | |||
| AK135775.1 | BAE22655.1 | |||
| AK145931.1 | BAE26762.1 | |||
| AK146313.1 | BAE27068.1 | |||
| AK160934.1 | BAE36099.1 | |||
| AK161819.1 | ||||
| AK198814.1 | ||||
| AK204129.1 | ||||
| BC048935.1 | AAH48935.1 |

Genomic Map