Assay Details
Target Gene Details
Entrez Gene ID: | 67998 |
Gene Name: | family with sequence similarity 134, member C |
Gene Aliases: |
1300010M03Rik, 4933404C01Rik, AI551748 |
Location: |
Chr.11:101096322-101119894 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam134c | NM_026501.2 | NP_080777.1 | ||
| NM_028933.3 | NP_083209.1 | |||
| AK004960.1 | BAB23700.1 | |||
| AK004979.1 | BAB23715.1 | |||
| AK016648.1 | ||||
| AK051025.1 | BAC34501.1 | |||
| AK151685.1 | BAE30609.1 | |||
| AK160000.1 | BAE35548.1 | |||
| AK163482.1 | BAE37364.1 | |||
| AK180187.1 | ||||
| BC006712.1 | AAH06712.1 | |||
| BC016089.1 | AAH16089.1 |

Genomic Map