Assay Details
Target Gene Details
Entrez Gene ID: | 23879 |
Gene Name: | fragile X mental retardation, autosomal homolog 2 |
Gene Aliases: |
Fxr2h |
Location: |
Chr.11:69632971-69653297 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxr2 | NM_011814.2 | NP_035944.2 | ||
| AB025269.1 | BAA82147.1 | |||
| AK154243.1 | BAE32459.1 | |||
| AK172042.1 | BAE42794.1 | |||
| AK203143.1 | ||||
| AK207619.1 | ||||
| BC057007.1 | AAH57007.1 | |||
| BC062971.1 | AAH62971.1 |
More Information
Set Membership: |
|

Genomic Map