Assay Details
Target Gene Details
Entrez Gene ID: | 107476 |
Gene Name: | acetyl-Coenzyme A carboxylase alpha |
Gene Aliases: |
A530025K05Rik, Acac, Acc1, Gm738 |
Location: |
Chr.11:84129677-84401651 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acaca | AJ619664.1 | CAF02251.1 | ||
| BY726813.1 |
Target Gene Details
Entrez Gene ID: | 628813 |
Gene Name: | predicted gene 11437 |
Gene Aliases: |
AI463690, OTTMUSG00000000997 |
Location: |
Chr.11:84148361-84167476 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11437 | NM_001037932.2 | NP_001033021.2 | ||
| AJ844653.1 | CAH59902.1 | |||
| BC147775.1 | ||||
| BC147791.1 |

Genomic Map