Assay Details
Target Gene Details
Entrez Gene ID: | 67480 |
Gene Name: | CWC25 spliceosome-associated protein homolog (S. cerevisiae) |
Gene Aliases: |
1300013D05Rik, AA473636, Ccdc49, R75228 |
Location: |
Chr.11:97745470-97766613 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cwc25 | NM_026186.4 | NP_080462.2 | ||
| AK004983.2 | BAB23718.1 | |||
| AK017849.1 | ||||
| AK035158.1 | ||||
| BC021478.1 | ||||
| BC079553.1 | AAH79553.1 |

Genomic Map