Assay Details
Target Gene Details
Entrez Gene ID: | 69125 |
Gene Name: | CCR4-NOT transcription complex, subunit 8 |
Gene Aliases: |
1500015I04Rik, 1810022F04Rik, AA536816, AU015770, AU043059 |
Location: |
Chr.11:58092799-58118596 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot8 | NM_026949.3 | 2 | 194 | NP_081225.1 |
| AK007581.1 | 2 | 258 | BAB25119.1 | |
| AK075727.1 | 2 | 164 | BAC35913.1 | |
| AK150757.1 | 2 | 275 | BAE29826.1 | |
| AK153907.1 | 2 | 197 | BAE32248.1 | |
| AY515608.1 | 1 | 161 | ||
| BC004040.1 | 2 | 194 | AAH04040.1 |
More Information
Set Membership: |
|

Genomic Map