Assay Details
Target Gene Details
Entrez Gene ID: | 103988 |
Gene Name: | glucokinase |
Gene Aliases: |
GLK, Gk, Gls006, HK4, HKIV, HXKP, Hlb62, MODY2 |
Location: |
Chr.11:5900816-5950081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gck | NM_001287386.1 | NP_001274315.1 | ||
| NM_010292.5 | NP_034422.2 | |||
| AK142698.1 | ||||
| BC011139.1 | AAH11139.1 | |||
| L38990.1 | AAB00360.1 | |||
| M58755.1 | AAA37703.1 |
More Information
Set Membership: |
|

Genomic Map