Assay Details
Target Gene Details
Entrez Gene ID: | 13494 |
Gene Name: | developmentally regulated GTP binding protein 1 |
Gene Aliases: |
AA408859, AI132520, Nedd3 |
Location: |
Chr.11:3249922-3266386 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Drg1 | NM_007879.1 | 6 | 733 | NP_031905.1 |
| AK044194.1 | 6 | 742 | ||
| AK080814.1 | 6 | 731 | BAC38030.1 | |
| AK144438.1 | 6 | 735 | BAE25890.1 | |
| AK155061.1 | 6 | 748 | BAE33021.1 | |
| AK184238.1 | 2 | 252 | ||
| AK188222.1 | 2 | 238 | ||
| BC039649.1 | 6 | 727 | AAH39649.1 | |
| D10715.1 | 6 | 733 | BAA01555.1 |

Genomic Map