Assay Details
Target Gene Details
Entrez Gene ID: | 208659 |
Gene Name: | family with sequence similarity 20, member A |
Gene Aliases: |
AI606893 |
Location: |
Chr.11:109669492-109722510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam20a | NM_153782.1 | NP_722477.1 | ||
| AK142932.1 | ||||
| BC029169.1 | AAH29169.1 |

Genomic Map