Assay Details
Target Gene Details
Entrez Gene ID: | 216613 |
Gene Name: | coiled-coil domain containing 85A |
Gene Aliases: |
- |
Location: |
Chr.11:28385684-28584441 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc85a | NM_001166661.2 | 4 | 2183 | NP_001160133.1 |
| NM_181577.5 | 5 | 2303 | NP_853555.2 | |
| AK045568.1 | BAC32419.1 | |||
| AK173298.1 | 4 | 2200 | BAD32576.1 | |
| BC020949.1 | 4 | 1266 | AAH20949.1 | |
| BC066065.1 | 5 | 1622 | AAH66065.1 |

Genomic Map