Assay Details
Target Gene Details
Entrez Gene ID: | 14252 |
Gene Name: | flotillin 2 |
Gene Aliases: |
AI573412, Esa, reggie-2 |
Location: |
Chr.11:78037868-78060434 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flot2 | NM_001040403.1 | NP_001035493.1 | ||
| NM_001284227.1 | NP_001271156.1 | |||
| NM_008028.2 | NP_032054.1 | |||
| AK004589.1 | BAB23392.1 | |||
| AK030199.1 | BAC26837.1 | |||
| AK149530.1 | BAE28942.1 | |||
| AK170557.1 | BAE41879.1 | |||
| AK186737.1 | ||||
| AK187814.1 | ||||
| AK200150.1 | ||||
| AK200750.1 | ||||
| AK201315.1 | ||||
| AK213895.1 | ||||
| BC070423.1 | AAH70423.1 | |||
| BY195482.1 | ||||
| BY204171.1 |

Genomic Map