Assay Details
Target Gene Details
Entrez Gene ID: | 18016 |
Gene Name: | neurofibromatosis 2 |
Gene Aliases: |
merlin |
Location: |
Chr.11:4757650-4849544 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf2 | NM_001252250.1 | 12 | 1828 | NP_001239179.1 |
| NM_001252251.1 | 12 | 1828 | NP_001239180.1 | |
| NM_001252252.1 | 12 | 1828 | NP_001239181.1 | |
| NM_001252253.1 | 12 | 1293 | NP_001239182.1 | |
| NM_010898.4 | 12 | 1828 | NP_035028.2 | |
| AK017343.1 | 12 | 1294 | BAB30698.1 | |
| AK045998.1 | 12 | 1650 | BAC32567.1 | |
| AK167685.1 | 9 | 919 | BAE39732.1 | |
| BB839081.1 | 3 | 357 | ||
| BC005442.1 | AAH05442.1 | |||
| BQ745455.1 | ||||
| L22987.1 | 12 | 1286 | ||
| L22988.1 | 12 | 1286 | ||
| L22989.1 | 12 | 1286 | ||
| L27090.1 | 12 | 1279 | AAA63648.1 | |
| L27105.1 | 12 | 1279 | AAA39807.1 | |
| L28176.1 | 12 | 1586 | AAA39808.1 | |
| X74671.1 | 12 | 1845 | CAA52737.1 | |
| X75759.1 | 1 | 72 | CAA53386.1 |
More Information
Set Membership: |
|

Genomic Map