Assay Details
Target Gene Details
Entrez Gene ID: | 16671 |
Gene Name: | keratin 33B |
Gene Aliases: |
Ha3, Ha4, Krt1-3, mHa3 |
Location: |
Chr.11:100023634-100029868 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt33b | NM_013570.1 | NP_038598.1 | ||
| AI466315.1 |

Genomic Map