Assay Details
Target Gene Details
Entrez Gene ID: | 16672 |
Gene Name: | keratin 34 |
Gene Aliases: |
4733401E01Rik, Ha2, Krt1-4 |
Location: |
Chr.11:100037351-100041554 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt34 | NM_027563.3 | NP_081839.1 | ||
| AK014631.1 | BAB29474.1 | |||
| BC125420.1 | ||||
| BC125446.1 |

Genomic Map