Assay Details
Target Gene Details
Entrez Gene ID: | 53617 |
Gene Name: | keratin 35 |
Gene Aliases: |
AI662400, Ha5, Krt1-24, Krtha5 |
Location: |
Chr.11:100092192-100096224 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt35 | NM_016880.2 | NP_058576.2 | ||
| AF020790.1 | AAD01693.1 | |||
| AK132593.1 | 4 | 1238 | ||
| BC100542.1 | AAI00543.1 | |||
| BC140965.1 |

Genomic Map