Assay Details
Target Gene Details
Entrez Gene ID: | 17308 |
Gene Name: | mannoside acetylglucosaminyltransferase 1 |
Gene Aliases: |
Mgat-1 |
Location: |
Chr.11:49244191-49263030 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat1 | NM_001110148.1 | NP_001103618.1 | ||
| NM_001110149.1 | NP_001103619.1 | |||
| NM_001110150.1 | NP_001103620.1 | |||
| NM_010794.3 | NP_034924.3 | |||
| AK004760.1 | BAB23541.1 | |||
| AK040121.1 | BAC30515.1 | |||
| AK087959.1 | BAC40058.1 | |||
| AK155322.1 | BAE33189.1 | |||
| BC031752.1 | AAH31752.1 | |||
| L07037.1 | AAA40478.1 | |||
| X77487.1 | ||||
| X77488.1 |
More Information
Set Membership: |
|

Genomic Map