Assay Details
Target Gene Details
Entrez Gene ID: | 17308 |
Gene Name: | mannoside acetylglucosaminyltransferase 1 |
Gene Aliases: |
Mgat-1 |
Location: |
Chr.11:49244191-49263030 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat1 | NM_001110148.1 | NP_001103618.1 | ||
| NM_010794.3 | NP_034924.3 | |||
| BC031752.1 | AAH31752.1 | |||
| BY088342.1 |
More Information
Set Membership: |
|

Genomic Map