Assay Details
Target Gene Details
Entrez Gene ID: | 544791 |
Gene Name: | myosin, heavy polypeptide 13, skeletal muscle |
Gene Aliases: |
MyHC-eo |
Location: |
Chr.11:67327103-67371502 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myh13 | NM_001081250.1 | NP_001074719.1 | ||
| AF336977.1 | AAL56215.1 |

Genomic Map