Assay Details
Target Gene Details
Entrez Gene ID: | 17896 |
Gene Name: | myosin, light polypeptide 4 |
Gene Aliases: |
ALC1, AMLC, ELC, ELC1a, GT1, MLC1a, Myla |
Location: |
Chr.11:104550530-104587219 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl4 | NM_010858.4 | NP_034988.2 | ||
| BC048485.1 | AAH48485.1 |
More Information
Set Membership: |
|

Genomic Map