Assay Details
Target Gene Details
Entrez Gene ID: | 18016 |
Gene Name: | neurofibromatosis 2 |
Gene Aliases: |
merlin |
Location: |
Chr.11:4757650-4849544 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf2 | NM_001252250.1 | NP_001239179.1 | ||
| NM_001252251.1 | NP_001239180.1 | |||
| NM_001252252.1 | NP_001239181.1 | |||
| NM_001252253.1 | NP_001239182.1 | |||
| NM_010898.4 | NP_035028.2 | |||
| AK017343.1 | BAB30698.1 | |||
| AK045998.1 | BAC32567.1 | |||
| AK167685.1 | BAE39732.1 | |||
| BC005442.1 | AAH05442.1 | |||
| DV654856.1 | ||||
| L22987.1 | ||||
| L22988.1 | ||||
| L22989.1 | ||||
| L27090.1 | AAA63648.1 | |||
| L27105.1 | AAA39807.1 | |||
| L28176.1 | AAA39808.1 | |||
| X74671.1 | CAA52737.1 |
More Information
Set Membership: |
|

Genomic Map