Assay Details
Target Gene Details
Entrez Gene ID: | 14923 |
Gene Name: | guanylate kinase 1 |
Gene Aliases: |
AA409738, AL033299, AL033300, AV026611, Gmk, Guk-1 |
Location: |
Chr.11:59183855-59192207 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Guk1 | NM_008193.3 | NP_032219.2 | ||
| AK004205.1 | BAB23219.2 | |||
| AK013514.1 | BAB28891.1 | |||
| BC024625.1 | AAH24625.1 | |||
| BY713424.1 | ||||
| U53514.1 | AAC52652.1 |
More Information
Set Membership: |
|

Genomic Map