Assay Details
Target Gene Details
Entrez Gene ID: | 17534 |
Gene Name: | mannose receptor, C type 2 |
Gene Aliases: |
Endo180, mKIAA0709, uPARAP |
Location: |
Chr.11:105292576-105351146 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrc2 | NM_008626.3 | 4 | 913 | NP_032652.3 |
| AK028157.1 | 4 | 904 | ||
| BC116642.1 | 4 | 839 | ||
| U56734.1 | 4 | 876 | AAC52729.1 |

Genomic Map