Assay Details
Target Gene Details
Entrez Gene ID: | 14723 |
Gene Name: | glycoprotein 1b, alpha polypeptide |
Gene Aliases: |
GPIba, GPIbalpha |
Location: |
Chr.11:70636651-70644030 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gp1ba | NM_010326.2 | 1 | 36 | NP_034456.2 |
| BC137765.1 |

Genomic Map