Assay Details
Target Gene Details
Entrez Gene ID: | 107656 |
Gene Name: | keratin 9 |
Gene Aliases: |
EPPK, K9, Krt1-9 |
Location: |
Chr.11:100186781-100193875 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt9 | NM_201255.2 | 3 | 785 | NP_957707.2 |
| AY497550.1 | 3 | 785 | AAR89518.1 |

Genomic Map