Assay Details
Target Gene Details
Entrez Gene ID: | 217325 |
Gene Name: | lethal giant larvae homolog 2 (Drosophila) |
Gene Aliases: |
9130006H11Rik, Llglh2 |
Location: |
Chr.11:115824029-115855780 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Llgl2 | NM_001252532.1 | NP_001239461.1 | ||
| NM_145438.2 | NP_663413.2 | |||
| AK143486.1 | ||||
| AK143798.1 | ||||
| AK167536.1 | BAE39604.1 | |||
| AY033650.1 | AAK52346.1 | |||
| BC037226.1 | AAH37226.1 |
More Information
Set Membership: |
|

Genomic Map