Assay Details
Target Gene Details
Entrez Gene ID: | 268510 |
Gene Name: | mannoside acetylglucosaminyltransferase 5, isoenzyme B |
Gene Aliases: |
C330018B01, GnT-IX, mGnTVB |
Location: |
Chr.11:116918856-116986944 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat5b | NM_172948.3 | NP_766536.2 | ||
| AB116028.1 | BAD08454.1 | |||
| AB119127.1 | BAC87707.1 | |||
| BC094604.1 | AAH94604.1 |
More Information
Set Membership: |
|

Genomic Map