Assay Details
Target Gene Details
Entrez Gene ID: | 56282 |
Gene Name: | mitochondrial ribosomal protein L12 |
Gene Aliases: |
0610034O11Rik, 1500031N16Rik, MRP-L12, Rpml12 |
Location: |
Chr.11:120484669-120488754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl12 | NM_027204.2 | NP_081480.2 | ||
| AK002757.1 | ||||
| AK005328.1 | BAB23955.1 | |||
| AK028168.1 | BAC25787.1 | |||
| AK134816.1 | BAE22297.1 | |||
| AK166969.1 | BAE39152.1 | |||
| AK168337.1 | BAE40275.1 | |||
| BC005712.1 | AAH05712.1 | |||
| BC019565.1 | AAH19565.1 |
More Information
Set Membership: |
|

Genomic Map