Assay Details
Target Gene Details
Entrez Gene ID: | 19062 |
Gene Name: | inositol polyphosphate 5-phosphatase K |
Gene Aliases: |
C62, Pps, SKIP |
Location: |
Chr.11:75630507-75648865 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5k | NM_008916.2 | 8 | 1048 | NP_032942.1 |
| AF483522.1 | 8 | 968 | AAL90796.1 | |
| AF483523.1 | 8 | 968 | AAL90797.1 | |
| AK052817.1 | 7 | 1792 | ||
| AK054436.1 | 8 | 1067 | BAC35778.1 | |
| AK078104.1 | 8 | 1059 | BAC37126.1 | |
| AK164290.1 | 8 | 1037 | BAE37720.1 | |
| BC014725.1 | 8 | 978 | ||
| BC066112.1 | 8 | 1048 | AAH66112.1 | |
| U96724.1 | 8 | 1079 | AAC53265.1 |
More Information
Set Membership: |
|

Genomic Map