Assay Details
Target Gene Details
Entrez Gene ID: | 17312 |
Gene Name: | C-type lectin domain family 10, member A |
Gene Aliases: |
CD301a, M-ASGP-BP-1, Mgl, Mgl1 |
Location: |
Chr.11:70166623-70170836 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec10a | NM_001204252.1 | NP_001191181.1 | ||
| NM_010796.3 | NP_034926.1 | |||
| AK140762.1 | ||||
| AK170779.1 | BAE42024.1 | |||
| BC014811.1 | AAH14811.1 | |||
| S36676.1 | AAB22171.1 |
More Information
Set Membership: |
|

Genomic Map