Assay Details
Target Gene Details
Entrez Gene ID: | 171283 |
Gene Name: | hepatitis A virus cellular receptor 1 |
Gene Aliases: |
AI503787, KIM-1, TIM-1, Tim1, Timd1 |
Location: |
Chr.11:46739823-46779578 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Havcr1 | NM_001166631.1 | NP_001160103.1 | ||
| NM_001166632.1 | NP_001160104.1 | |||
| NM_134248.2 | NP_599009.2 | |||
| AF399829.1 | AAL35774.1 | |||
| AF399830.1 | AAL35775.1 | |||
| AK133466.1 | BAE21672.1 | |||
| BC053400.1 | AAH53400.1 |
More Information
Set Membership: |
|

Genomic Map