Assay Details
Target Gene Details
Entrez Gene ID: | 73158 |
Gene Name: | La ribonucleoprotein domain family, member 1 |
Gene Aliases: |
1810024J12Rik, 3110040D16Rik, Larp, mKIAA0731 |
Location: |
Chr.11:58009064-58062034 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Larp1 | NM_028451.1 | NP_082727.1 | ||
| AK129202.1 | BAC98012.1 |
More Information
Set Membership: |
|

Genomic Map