Assay Details
Target Gene Details
Entrez Gene ID: | 100042498 |
Gene Name: | Mir22 host gene (non-protein coding) |
Gene Aliases: |
2210403K04Rik, mir-22 |
Location: |
Chr.11:75461539-75466690 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir22hg | NR_030711.1 | |||
| AK008813.1 | ||||
| AK018628.1 | ||||
| AK021103.1 |

Genomic Map