Assay Details
Target Gene Details
Entrez Gene ID: | 216864 |
Gene Name: | macrophage galactose N-acetyl-galactosamine specific lectin 2 |
Gene Aliases: |
CD301b |
Location: |
Chr.11:70130125-70137542 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgl2 | NM_145137.2 | NP_660119.1 | ||
| AK085751.1 | 8 | 764 | BAC39530.1 | |
| AY103461.1 | AAM52097.1 | |||
| BC106833.1 | AAI06834.1 | |||
| BC110475.2 | AAI10476.1 | |||
| BC111520.1 | AAI11521.1 | |||
| EU010011.1 | 9 | 839 |
More Information
Set Membership: |
|

Genomic Map