Assay Details
Target Gene Details
Entrez Gene ID: | 17966 |
Gene Name: | neighbor of Brca1 gene 1 |
Gene Aliases: |
mKIAA0049 |
Location: |
Chr.11:101552107-101581951 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbr1 | NM_001252220.1 | 23 | 3415 | NP_001239149.1 |
| NM_001252222.1 | 22 | 3127 | NP_001239151.1 | |
| NM_001252223.1 | NP_001239152.1 | |||
| NM_008676.3 | 23 | 3238 | NP_032702.1 | |
| AF227188.1 | 23 | 3367 | AAF74118.1 | |
| AF408436.1 | 13 | 1962 | AAL11728.1 | |
| AK077479.1 | 14 | 1915 | ||
| AK128992.1 | 22 | 2919 | ||
| AK129043.1 | BAC97853.1 | |||
| AK154081.1 | ||||
| BC052709.1 | 22 | 2970 | ||
| BC129867.1 | 22 | 2890 | ||
| BC129868.1 | 22 | 2890 | ||
| U73039.1 | 23 | 3034 | AAC53025.1 |

Genomic Map