Assay Details
Target Gene Details
Entrez Gene ID: | 17966 |
Gene Name: | neighbor of Brca1 gene 1 |
Gene Aliases: |
mKIAA0049 |
Location: |
Chr.11:101552107-101581951 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbr1 | NM_001252220.1 | NP_001239149.1 | ||
| NM_001252222.1 | NP_001239151.1 | |||
| NM_001252223.1 | NP_001239152.1 | |||
| NM_008676.3 | NP_032702.1 | |||
| AF227188.1 | AAF74118.1 | |||
| AK128992.1 | ||||
| AK129043.1 | BAC97853.1 | |||
| AK154081.1 | ||||
| BC052709.1 | ||||
| BC129867.1 | ||||
| BC129868.1 | ||||
| CO433793.1 | ||||
| CV558786.1 | ||||
| U73039.1 | AAC53025.1 |

Genomic Map