Assay Details
Target Gene Details
Entrez Gene ID: | 56310 |
Gene Name: | G protein pathway suppressor 2 |
Gene Aliases: |
AI505953 |
Location: |
Chr.11:69913980-69917150 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gps2 | NM_019726.3 | NP_062700.2 | ||
| AF153696.1 | AAD38393.1 | |||
| AK185728.1 | ||||
| BC011317.1 | AAH11317.1 | |||
| BC138879.1 | ||||
| BC138880.1 |
More Information
Set Membership: |
|

Genomic Map