Assay Details
Target Gene Details
Entrez Gene ID: | 171285 |
Gene Name: | hepatitis A virus cellular receptor 2 |
Gene Aliases: |
TIM-3, Tim3, Timd3 |
Location: |
Chr.11:46454547-46481255 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Havcr2 | NM_134250.2 | NP_599011.2 | ||
| AF399831.1 | AAL35776.1 | |||
| AF450241.1 | AAL65156.1 | |||
| AK148474.1 | BAE28574.1 | |||
| AK170775.1 | ||||
| AY553334.1 | AAS59531.1 | |||
| BC106851.2 | AAI06852.1 | |||
| BC106852.2 | AAI06853.1 |
More Information
Set Membership: |
|

Genomic Map