Assay Details
Target Gene Details
Entrez Gene ID: | 59013 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H1 |
Gene Aliases: |
AI642080, E430005G16Rik, Hnrnph, Hnrph1 |
Location: |
Chr.11:50377694-50386528 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnph1 | NM_021510.2 | NP_067485.1 | ||
| AK052983.1 | ||||
| AK088169.1 | BAC40188.1 | |||
| AK165283.1 | BAE38120.1 | |||
| BC042187.1 | AAH42187.2 | |||
| BC056224.1 | AAH56224.1 | |||
| CO428102.1 | ||||
| Y14196.1 | CAA74583.1 |
More Information
Set Membership: |
|

Genomic Map