Assay Details
Target Gene Details
Entrez Gene ID: | 16211 |
Gene Name: | karyopherin (importin) beta 1 |
Gene Aliases: |
AA409963, IPOB, Impnb |
Location: |
Chr.11:97159710-97187892 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kpnb1 | NM_008379.3 | 4 | 733 | NP_032405.3 |
| AK077709.1 | 4 | 723 | ||
| AK147175.1 | 4 | 752 | BAE27738.1 | |
| AK169171.1 | 4 | 703 | BAE40950.1 | |
| BC052438.1 | 4 | 488 | AAH52438.1 | |
| BC052711.1 | 4 | 521 | AAH52711.1 | |
| BC055115.1 | 4 | 718 | AAH55115.1 | |
| D45836.1 | 4 | 624 | BAA08273.1 | |
| D67015.1 | 4 | 461 | BAA11034.1 |
More Information
Set Membership: |
|

Genomic Map