Assay Details
Target Gene Details
Entrez Gene ID: | 194952 |
Gene Name: | jumonji domain containing 4 |
Gene Aliases: |
6430559I23 |
Location: |
Chr.11:59450045-59458567 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmjd4 | NM_001205068.1 | NP_001191997.1 | ||
| NM_178659.6 | NP_848774.1 | |||
| NR_037992.1 | ||||
| AK032477.1 | ||||
| AK037954.1 | BAC29907.1 | |||
| AK038650.1 | ||||
| AK039116.1 | ||||
| AK047586.1 | ||||
| AK052144.1 | BAC34856.1 | |||
| AK079104.1 | BAC37541.1 | |||
| AK084669.1 | ||||
| AK145394.1 | BAE26409.1 | |||
| AK171919.1 | ||||
| BC049148.1 | AAH49148.1 | |||
| BC116707.1 | ||||
| BC118020.1 |
More Information
Set Membership: |
|

Genomic Map