Assay Details
Target Gene Details
Entrez Gene ID: | 83431 |
Gene Name: | nuclear distribution gene E-like homolog 1 (A. nidulans) |
Gene Aliases: |
2600006O07Rik, MITAP1, NUDEL, mNudel |
Location: |
Chr.11:68821434-68853131 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndel1 | NM_023668.2 | 3 | 366 | NP_076157.2 |
| AF290472.1 | 3 | 298 | AAG10061.1 | |
| AF323918.1 | 3 | 386 | AAG42496.1 | |
| AK011168.1 | 3 | 276 | BAB27443.1 | |
| AK146029.1 | 3 | 295 | ||
| AK212133.1 | 2 | 145 | ||
| BC046796.1 | 3 | 297 | AAH46796.1 | |
| CJ092278.1 | 3 | 367 |

Genomic Map