Assay Details
Target Gene Details
Entrez Gene ID: | 14786 |
Gene Name: | growth factor receptor bound protein 7 |
Gene Aliases: |
mKIAA4028 |
Location: |
Chr.11:98446834-98455373 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grb7 | NM_010346.2 | NP_034476.1 | ||
| AK057640.1 | ||||
| AK145996.1 | BAE26817.1 | |||
| AK168438.1 | BAE40343.1 | |||
| AK220277.1 | BAD90202.1 | |||
| BC003295.1 | AAH03295.1 | |||
| M94450.1 | AAA37733.1 |
More Information
Set Membership: |
|

Genomic Map