Assay Details
Target Gene Details
Entrez Gene ID: | 15485 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 1 |
Gene Aliases: |
17HSDB1, 17beta-HSD, E2DH, Hsd17ba |
Location: |
Chr.11:101077968-101081188 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd17b1 | NM_010475.1 | NP_034605.1 | ||
| AK019406.1 | BAB31706.1 | |||
| AK143346.1 | BAE25350.1 | |||
| BC125657.1 | ||||
| BC125659.1 | ||||
| X89627.1 | CAA61770.1 |
More Information
Set Membership: |
|

Genomic Map