Assay Details
Target Gene Details
Entrez Gene ID: | 16663 |
Gene Name: | keratin 13 |
Gene Aliases: |
CK13, K13, Krt-1.13, Krt1-13 |
Location: |
Chr.11:100117596-100121495 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt13 | NM_010662.1 | NP_034792.1 | ||
| AK004635.1 | ||||
| AK009677.1 | ||||
| AK010156.1 | ||||
| AK132640.1 | BAE21276.1 | |||
| BC060286.1 | AAH60286.1 | |||
| X03492.1 | CAA27208.1 |
More Information
Set Membership: |
|

Genomic Map