Assay Details
Target Gene Details
Entrez Gene ID: | 16664 |
Gene Name: | keratin 14 |
Gene Aliases: |
AI626930, K14, Krt-1.14, Krt1-14 |
Location: |
Chr.11:100203162-100207510 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt14 | NM_016958.1 | NP_058654.1 | ||
| BC003325.1 | AAH03325.1 | |||
| BC011074.1 | AAH11074.1 | |||
| M13806.1 | AAA39392.1 |
More Information
Set Membership: |
|

Genomic Map