Assay Details
Target Gene Details
Entrez Gene ID: | 83431 |
Gene Name: | nuclear distribution gene E-like homolog 1 (A. nidulans) |
Gene Aliases: |
2600006O07Rik, MITAP1, NUDEL, mNudel |
Location: |
Chr.11:68821434-68853131 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndel1 | NM_023668.2 | NP_076157.2 | ||
| AF290472.1 | AAG10061.1 | |||
| AF323918.1 | AAG42496.1 | |||
| AK011168.1 | BAB27443.1 | |||
| AK146029.1 | ||||
| BC046796.1 | AAH46796.1 |

Genomic Map